4-(Benzyloxy)-3-ethoxybenzaldehyde ≥98%

Supplier: Matrix Scientific

006543-5G 006543-25G
101790-692EA 67.69 USD
101790-692 101790-694
4-(Benzyloxy)-3-ethoxybenzaldehyde ≥98%
4-(Benzyloxy)-3-ethoxybenzaldehyde

Hazard: Irritant

Formula: C₁₆H₁₆O₃
Melting Pt: 56…58 °C
MDL Number: MFCD00615145
CAS Number: 60186-33-6

Order Now

Specification Test Results

Melting point 56-58°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR