Tetrabutylammonium fluoride trihydrate ≥97%

Supplier: Matrix Scientific

006516-100G 006516-500G
101790-678EA 89.23 USD
101790-678 101790-680
Tetrabutylammonium fluoride trihydrate ≥97%
Tetrabutylammonium fluoride trihydrate

TCA listed: T
Hazard: Irritant/Irritant-Harmful

Formula: C₁₆H₃₆FN·3H₂O
MW: 315.51 g/mol
Melting Pt: 62…63 °C
MDL Number: MFCD00149981
CAS Number: 87749-50-6
UN: 2811
ADR: 6.1 II

Order Now

Specification Test Results

Melting point 62-63°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR