N-Pentafluorophenyldichloromaleimide ≥97%

Supplier: Matrix Scientific

006502-1G 006502-5G
101790-656EA 53.85 USD
101790-656 101790-658
N-Pentafluorophenyldichloromaleimide ≥97%
N-Pentafluorophenyldichloromaleimide

Hazard: Irritant

Formula: C₁₀Cl₂F₅NO₂
Melting Pt: 148…151 °C
MDL Number: MFCD00054728
CAS Number: 186958-58-7

Order Now

Specification Test Results

Melting point 148-151°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR