Bis(4-acetylphenyl)methane ≥98%

Supplier: Matrix Scientific

006408-1G 006408-5G
101790-526EA 27.69 USD
101790-526 101790-528
Bis(4-acetylphenyl)methane ≥98%
Bis(4-acetylphenyl)methane

Hazard: Irritant

Formula: C₁₇H₁₆O₂
Melting Pt: 93 °C
MDL Number: MFCD00053708
CAS Number: 790-82-9

Order Now

Specification Test Results

Melting point 93°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR