3,3'-Bis(trifluoromethyl)benzidine ≥97%

Supplier: Matrix Scientific

006379-250MG 006379-1G
101790-464EA 118.46 USD
101790-464 101790-466
3,3'-Bis(trifluoromethyl)benzidine ≥97%
3,3'-Bis(trifluoromethyl)benzidine

Hazard: Irritant-Harmful

Formula: C₁₄H₁₀F₆N₂
Melting Pt: 180…181 °C
MDL Number: MFCD00236672
CAS Number: 346-88-3

Order Now

Specification Test Results

Melting point 180-181°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR