Methyl p-Tolyl sulfone ≥97%

Supplier: Matrix Scientific

Synonyms: 4-(Methylsulphonyl)toluene, 1-Methyl-4-(methylsulphonyl)benzene

006346-5G 006346-25G
101790-398EA 15.38 USD
101790-398 101790-400
Methyl p-Tolyl sulfone ≥97%
Methyl p-Tolyl sulfone

Hazard: Irritant

Formula: C₈H₁₀O₂S
MW: 170.23 g/mol
Melting Pt: 87…88 °C
MDL Number: MFCD00014742
CAS Number: 3185-99-7

Order Now

Specification Test Results

Melting point 87-88°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR