4,4'-Dimethyloctafluorobiphenyl ≥97%

Supplier: Matrix Scientific

006343-1G 006343-5G
101790-394EA 95.38 USD
101790-394 101790-396
4,4'-Dimethyloctafluorobiphenyl ≥97%
4,4'-Dimethyloctafluorobiphenyl

Hazard: Irritant

Formula: C₁₄H₆F₈
Melting Pt: 148…149 °C
MDL Number: MFCD00000314
CAS Number: 26475-18-3

Order Now

Specification Test Results

Melting point 148-149°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR