2-(Methylthio)nicotinic acid hydrazide
Supplier: Matrix Scientific
006310-1G
006310-5G
101790-370EA
43.08
USD
101790-370
101790-372
2-(Methylthio)nicotinic acid hydrazide
2-(Methylthio)nicotinic acid hydrazide
Hazard: Irritant
Formula:
C₇H₉N₃OS Melting Pt: 142-144°C |
CAS Number:
232926-33-9 |
Specification Test Results
Melting point | 142-144°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...