4-(Methylthio)benzhydrazide

Supplier: Matrix Scientific

006302-1G 006302-5G
101790-358EA 70.77 USD
101790-358 101790-360
4-(Methylthio)benzhydrazide
4-(Methylthio)benzhydrazide

Hazard: Irritant

Formula: C₈H₁₀N₂OS
Melting Pt: 161-163°C
MDL Number: MFCD01310830
CAS Number: 81104-42-9

Order Now

Specification Test Results

Melting point 161-163°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR