2,2',3,3',5,5',6,6'-Octafluorobiphenyl ≥97%

Supplier: Matrix Scientific

006262-5G 006262-25G
101790-292EA 144.62 USD
101790-292 101790-294
2,2',3,3',5,5',6,6'-Octafluorobiphenyl ≥97%
2,2',3,3',5,5',6,6'-Octafluorobiphenyl

Hazard: Irritant

Formula: C₁₂H₂F₈
Melting Pt: 84…86 °C
MDL Number: MFCD00000312
CAS Number: 3883-86-1

Order Now

Specification Test Results

Melting point 84-86°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR