Chloropentafluoroacetone monohydrate ≥97%

Supplier: Matrix Scientific

006257-5G 006257-25G
101790-278EA 76.92 USD
101790-278 101790-280
Chloropentafluoroacetone monohydrate ≥97%
Chloropentafluoroacetone monohydrate

Hazard: Toxic

Formula: C₃ClF₅O·H₂O
MW: 200.49 g/mol
Melting Pt: 26.0…26.5 °C
MDL Number: MFCD00150647
CAS Number: 6984-99-2

Order Now

Specification Test Results

Melting point 26.0-26.5°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR