1-[3,5-Bis(trifluoromethyl)phenyl]pyrrole

Supplier: Matrix Scientific

006177-1G 006177-5G
101790-198EA 35.38 USD
101790-198 101790-200
1-[3,5-Bis(trifluoromethyl)phenyl]pyrrole
1-[3,5-Bis(trifluoromethyl)phenyl]pyrrole

Hazard: Irritant

Formula: C₁₂H₇F₆N
MW: 279.19
Melting Pt: 43°C
MDL Number: MFCD00053048
CAS Number: 175136-60-4

Order Now

Specification Test Results

Melting point 43°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR