3-[3,5-Bis(trifluoromethyl)phenyl]acrylonitrile

Supplier: Matrix Scientific

006176-1G 006176-5G
101790-194EA 1163.08 USD
101790-194 101790-196
3-[3,5-Bis(trifluoromethyl)phenyl]acrylonitrile
3-[3,5-Bis(trifluoromethyl)phenyl]acrylonitrile

Hazard: Irritant

Formula: C₁₁H₅F₆N
Melting Pt: 80 °C
MDL Number: MFCD00051605
CAS Number: 175136-63-7

Order Now

Specification Test Results

Melting point 80°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR