N-[3,5-Bis(trifluoromethyl)benzenesulfonyl]-L-methionine methyl ester

Supplier: Matrix Scientific

Synonyms: Methyl (2S)-2-({[3,5-bis(trifluoromethyl)phenyl]sulphonyl}amino)-4-(methylthio)butanoate

006174-1G 006174-5G
101790-190EA 38.46 USD
101790-190 101790-192
N-[3,5-Bis(trifluoromethyl)benzenesulfonyl]-L-methionine methyl ester
N-[3,5-Bis(trifluoromethyl)benzenesulfonyl]-L-methionine methyl ester

Hazard: Irritant

Formula: C₁₄H₁₅F₆NO₄S₂
MW: 439.4 g/mol
Melting Pt: 99…102 °C
MDL Number: MFCD00084899
CAS Number: 175202-21-8

Order Now

Specification Test Results

Melting point 99-102°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR