1-(4-Fluorophenyl)imidazoline-2-thione

Supplier: Matrix Scientific

Synonyms: 1-(4-Fluorophenyl)-1H-imidazole-2(3H)-thione

006097-1G 006097-5G
101790-066EA 1030.77 USD
101790-066 101790-068
1-(4-Fluorophenyl)imidazoline-2-thione
1-(4-Fluorophenyl)imidazoline-2-thione

Hazard: Irritant

Formula: C₉H₇FN₂S
MW: 194.23 g/mol
Melting Pt: 207…209 °C
MDL Number: MFCD00060483
CAS Number: 17452-07-2

Order Now

Specification Test Results

Melting point 207-209°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR