1-(3-Fluorophenyl)imidazoline-2-thione

Supplier: Matrix Scientific

006096-1G 006096-5G
101790-062EA 46.15 USD
101790-062 101790-064
1-(3-Fluorophenyl)imidazoline-2-thione
1-(3-Fluorophenyl)imidazoline-2-thione

Hazard: Irritant

Formula: C₉H₇FN₂S
Melting Pt: 162…164 °C
MDL Number: MFCD00060479
CAS Number: 17452-26-5

Order Now

Specification Test Results

Melting point 162-164°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR