1-(3-Chlorophenyl)imidazoline-2-thione ≥98%

Supplier: Matrix Scientific

006094-1G 006094-5G
101790-054EA 36.92 USD
101790-054 101790-056
1-(3-Chlorophenyl)imidazoline-2-thione ≥98%
1-(3-Chlorophenyl)imidazoline-2-thione

Hazard: Irritant

Formula: C₉H₇ClN₂S
Melting Pt: 161…163 °C
MDL Number: MFCD00060480
CAS Number: 30192-81-5

Order Now

Specification Test Results

Melting point 161-163°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR