1-(2-Chlorophenyl)imidazoline-2-thione ≥98%

Supplier: Matrix Scientific

006093-1G 006093-5G
101790-050EA 38.46 USD
101790-050 101790-052
1-(2-Chlorophenyl)imidazoline-2-thione ≥98%
1-(2-Chlorophenyl)imidazoline-2-thione

Hazard: Irritant

Formula: C₉H₇ClN₂S
Melting Pt: 259…261 °C
MDL Number: MFCD00041200
CAS Number: 51581-47-6

Order Now

Specification Test Results

Melting point 259-261°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR