2-(4-Chlorophenyl)-3-(trifluoromethyl)pyrazole-4-carboxylic acid hydrazide

Supplier: Matrix Scientific

006073-1G 006073-5G
101790-018EA 90.77 USD
101790-018 101790-020
2-(4-Chlorophenyl)-3-(trifluoromethyl)pyrazole-4-carboxylic acid hydrazide
2-(4-Chlorophenyl)-3-(trifluoromethyl)pyrazole-4-carboxylic acid hydrazide

Hazard: Irritant

Formula: C₁₁H₈ClF₃N₄O
Melting Pt: 179…181 °C
MDL Number: MFCD00068154
CAS Number: 175137-34-5

Order Now

Specification Test Results

Melting point 179-181°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR