2-(4-Chlorophenyl)-3-(trifluoromethyl)pyrazole-4-carboxylic acid

Supplier: Matrix Scientific

006072-1G 006072-5G
101790-014EA 27.69 USD
101790-014 101790-016
2-(4-Chlorophenyl)-3-(trifluoromethyl)pyrazole-4-carboxylic acid
2-(4-Chlorophenyl)-3-(trifluoromethyl)pyrazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₆ClF₃N₂O₂
MW: 290.63 g/mol
Melting Pt: 160…164 °C
MDL Number: MFCD00068142
CAS Number: 98534-80-6

Order Now

Specification Test Results

Melting point 160-164°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR