5-(4-Chlorophenyl)-2-(trifluoromethyl)furan-3-carbonyl chloride ≥97%

Supplier: Matrix Scientific

006070-250MG 006070-1G
101790-006EA 46.15 USD
101790-006 101790-008
5-(4-Chlorophenyl)-2-(trifluoromethyl)furan-3-carbonyl chloride ≥97%
5-(4-Chlorophenyl)-2-(trifluoromethyl)furan-3-carbonyl chloride

Hazard: Irritant

Formula: C₁₂H₅Cl₂F₃O₂
Melting Pt: 135…137 °C
MDL Number: MFCD00275554
CAS Number: 175276-61-6

Order Now

Specification Test Results

Melting point 135-137°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR