4'-(4-Fluorobenzyloxy)-3'-nitroacetophenone

Supplier: Matrix Scientific

005963-250MG 005963-1G
101789-860EA 44.62 USD
101789-860 101789-862
4'-(4-Fluorobenzyloxy)-3'-nitroacetophenone
4'-(4-Fluorobenzyloxy)-3'-nitroacetophenone

Hazard: Irritant

Formula: C₁₅H₁₂FNO₄
Melting Pt: 105-107°C
MDL Number: MFCD00173872
CAS Number: 175136-24-0

Order Now

Specification Test Results

Melting point 105-107°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR