5-(4-Chlorobenzyloxycarbonyl)-4-(trifluoromethyl)pyrimidin-2-yl hydrazine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Chlorobenzyl 2-hydrazino-4-(trifluoromethyl)pyrimidine-5-carboxylate

005960-250MG 005960-1G
101789-854EA 158.46 USD
101789-854 101789-856
5-(4-Chlorobenzyloxycarbonyl)-4-(trifluoromethyl)pyrimidin-2-yl hydrazine ≥95%
5-(4-Chlorobenzyloxycarbonyl)-4-(trifluoromethyl)pyrimidin-2-yl hydrazine

Hazard: Irritant

Formula: C₁₃H₁₀ClF₃N₄O₂
MW: 346.7 g/mol
Melting Pt: 111…113 °C
MDL Number: MFCD00068153
CAS Number: 175137-33-4

Order Now

Specification Test Results

Melting point 111-113°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR