1-(4-Fluorobenzyl)-4-(2-hydroxyethyl)piperazine

Supplier: Matrix Scientific

005959-1G 005959-5G
101789-850EA 72.31 USD
101789-850 101789-852
1-(4-Fluorobenzyl)-4-(2-hydroxyethyl)piperazine
1-(4-Fluorobenzyl)-4-(2-hydroxyethyl)piperazine

Hazard: Irritant

Formula: C₁₃H₁₉FN₂O
Melting Pt: 55…56 °C
MDL Number: MFCD00203973
CAS Number: 174561-11-6

Order Now

Specification Test Results

Melting point 55-56°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR