4-Trifluoroacetyl-3-methyl-1-phenyl-5-pyrazolone
Supplier: Matrix Scientific
Synonyms:
5-Methyl-2-phenyl-4-(2,2,2-trifluoroacetyl)-1H-pyrazol-3(2H)-one, 3-Methyl-1-phenyl-4-trifluoroacetyl-5-pyrazolone
005935-1G
005935-5G
101789-790EA
84.62
USD
101789-790
101789-792
4-Trifluoroacetyl-3-methyl-1-phenyl-5-pyrazolone
4-Trifluoroacetyl-3-methyl-1-phenyl-5-pyrazolone
Hazard: Irritant
Formula:
C₁₂H₉F₃N₂O₂ MW: 270.21 g/mol Melting Pt: 143 °C |
MDL Number:
MFCD00143378 CAS Number: 1691-93-6 |
Specification Test Results
Melting point | 143°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...