1-(2-Chloro-6-fluorophenyl)-2-nitroethene

Supplier: Matrix Scientific

005930-1G 005930-5G
101789-778EA 60 USD
101789-778 101789-780
1-(2-Chloro-6-fluorophenyl)-2-nitroethene
1-(2-Chloro-6-fluorophenyl)-2-nitroethene

Hazard: Irritant

Formula: C₈H₅ClFNO₂
Melting Pt: 63 °C
MDL Number: MFCD00053064
CAS Number: 60610-68-6

Order Now

Specification Test Results

Melting point 63°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR