3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carbonitrile

Supplier: Matrix Scientific

005927-1G 005927-5G
101789-774EA 73.85 USD
101789-774 101789-776
3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carbonitrile
3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carbonitrile

Hazard: Irritant

Formula: C₁₁H₆ClFN₂O
Melting Pt: 85°C
MDL Number: MFCD00052551
CAS Number: 175204-41-8

Order Now

Specification Test Results

Melting point 85°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR