4-[(2-Chloro-6-fluorobenzyl)oxy]benzaldehyde

Supplier: Matrix Scientific

005924-1G 005924-5G
101789-770EA 93.85 USD
101789-770 101789-772
4-[(2-Chloro-6-fluorobenzyl)oxy]benzaldehyde
4-[(2-Chloro-6-fluorobenzyl)oxy]benzaldehyde

Hazard: Irritant

Formula: C₁₄H₁₀ClFO₂
Melting Pt: 78-80°C
MDL Number: MFCD00052227
CAS Number: 172932-10-4

Order Now

Specification Test Results

Melting point 78-80°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR