2-(2-Chloro-6-fluorobenzyl)-4,5-dichloropyridazine-3-(2H)-one

Supplier: Matrix Scientific

005922-1G 005922-5G
101789-766EA 87.69 USD
101789-766 101789-768
2-(2-Chloro-6-fluorobenzyl)-4,5-dichloropyridazine-3-(2H)-one
2-(2-Chloro-6-fluorobenzyl)-4,5-dichloropyridazine-3-(2H)-one

Hazard: Irritant

Formula: C₁₁H₆Cl₃FN₂O
Melting Pt: 137…138 °C
MDL Number: MFCD00067774
CAS Number: 175135-45-2

Order Now

Specification Test Results

Melting point 137-138°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR