3-(3-Fluoro-4-methoxybenzoyl)propionic acid ≥98%

Supplier: Matrix Scientific

005915-1G 005915-5G
101789-748EA 36.92 USD
101789-748 101789-750
3-(3-Fluoro-4-methoxybenzoyl)propionic acid ≥98%
3-(3-Fluoro-4-methoxybenzoyl)propionic acid

Purity: 98% CAS: 347-63-7 MDL Number: MFCD00020538 Molecular Formula: C11H11FO4 Molecular Weight: 226,21 Hazard: Irritant

Formula: C₁₁H₁₁FO₄
Melting Pt: 171-173°C
MDL Number: MFCD00020538
CAS Number: 347-63-7

Order Now

Specification Test Results

Melting point 171-173°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR