4'-Chloro-3'-methyl-2-nitro-4-(trifluoromethyl)diphenyl ether

Supplier: Matrix Scientific

005909-1G 005909-5G
101789-736EA 52.31 USD
101789-736 101789-738
4'-Chloro-3'-methyl-2-nitro-4-(trifluoromethyl)diphenyl ether
4'-Chloro-3'-methyl-2-nitro-4-(trifluoromethyl)diphenyl ether

Hazard: Irritant

Formula: C₁₄H₉ClF₃NO₃
MW: 331.67 g/mol
Melting Pt: 57…59 °C
MDL Number: MFCD00052259
CAS Number: 1799-97-9

Order Now

Specification Test Results

Melting point 57-59°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR