S-[5-Chloro-3-(trifluoromethyl)pyridin-2-yl]-S-methylsulfilimine-2,4,6-trimethylbenzenesulfonate

Supplier: Matrix Scientific

005907-250MG 005907-1G
101789-728EA 56.92 USD
101789-728 101789-730
S-[5-Chloro-3-(trifluoromethyl)pyridin-2-yl]-S-methylsulfilimine-2,4,6-trimethylbenzenesulfonate
S-[5-Chloro-3-(trifluoromethyl)pyridin-2-yl]-S-methylsulfilimine-2,4,6-trimethylbenzenesulfonate

Hazard: Irritant

Formula: C₁₆H₁₈ClF₃N₂O₃S₂
MW: 442.91 g/mol
Melting Pt: 167…170 °C
MDL Number: MFCD00068013
CAS Number: 175203-07-3

Order Now

Specification Test Results

Melting point 167-170°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR