3,5-Bis(trifluoromethyl)benzhydrazide ≥97%

Supplier: Matrix Scientific

Synonyms: 3,5-Bis(trifluoromethyl)benzohydrazide

005859-1G 005859-5G
101789-664EA 58.46 USD
101789-664 101789-666
3,5-Bis(trifluoromethyl)benzhydrazide ≥97%
3,5-Bis(trifluoromethyl)benzhydrazide

Hazard: Irritant

Formula: C₉H₆F₆N₂O
MW: 272.15 g/mol
Melting Pt: 132-134°C
MDL Number: MFCD00051848
CAS Number: 26107-82-4

Order Now

Specification Test Results

Melting point 132-134°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR