3,5-Bis(trifluoromethyl)phenylhydrazine hydrochloride ≥98%

Supplier: Matrix Scientific

005834-1G 005834-5G
101789-606EA 27.69 USD
101789-606 101789-608
3,5-Bis(trifluoromethyl)phenylhydrazine hydrochloride ≥98%
3,5-Bis(trifluoromethyl)phenylhydrazine hydrochloride

Hazard: Irritant

Formula: C₈H₇ClF₆N₂
MW: 280.60 g/mol
Melting Pt: 300°C
MDL Number: MFCD03094165
CAS Number: 502496-23-3

Order Now

Specification Test Results

Melting point 80-83°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR