4-Carboxyphenylboronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Boronobenzoic Acid, Ark Pharm, 4-Carboxybenzeneboronic acid

005716-5G 005716-25G
101789-420EA 29.23 USD
101789-420 101789-422
4-Carboxyphenylboronic acid ≥95%
4-Carboxyphenylboronic acid

Hazard: Irritant/Store in Freezer

Formula: C₇H₇BO₄
MW: 165.94 g/mol
Melting Pt: 238-240°C
MDL Number: MFCD00151801
CAS Number: 14047-29-1

Order Now

Specification Test Results

Melting point 238-240°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR