3-Phenylbenzonitrile

Supplier: Matrix Scientific

Synonyms: [1,1'-biphenyl]-3-carbonitrile

005698-1G 005698-5G
101789-382EA 152.31 USD
101789-382 101789-384
3-Phenylbenzonitrile
3-Phenylbenzonitrile

Hazard: Irritant

Formula: C₁₃H₉N
MW: 179.2173 g/mol
Melting Pt: 44…46 °C
MDL Number: MFCD00236364
CAS Number: 24973-50-0

Order Now

Specification Test Results

Melting point 44-46°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR