1-(4-Methoxyphenyl)imidazoline-2-thione ≥98%

Supplier: Matrix Scientific

005664-1G 005664-5G
101789-306EA 38.46 USD
101789-306 101789-308
1-(4-Methoxyphenyl)imidazoline-2-thione ≥98%
1-(4-Methoxyphenyl)imidazoline-2-thione

Hazard: Irritant

Formula: C₁₀H₁₀N₂OS
Melting Pt: 216…217 °C
MDL Number: MFCD00041376
CAS Number: 17452-14-1

Order Now

Specification Test Results

Melting point 216-217°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR