6,7-Dichloro-2-methylquinoline

Supplier: Matrix Scientific

005612-1G 005612-5G
101789-238EA 176.92 USD
101789-238 101789-240
6,7-Dichloro-2-methylquinoline
6,7-Dichloro-2-methylquinoline

Hazard: Irritant

Formula: C₁₀H₇Cl₂N
Melting Pt: 121…123 °C
MDL Number: MFCD00270103
CAS Number: 71063-12-2

Order Now

Specification Test Results

Melting point 121-123°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR