4-(Trifluoromethylthio)benzaldehyde ≥98%

Supplier: Matrix Scientific

Synonyms: 4-((Trifluoromethyl)thio)benzaldehyde

005520-1G 005520-5G
101789-080EA 15.38 USD
101789-080 101789-082
4-(Trifluoromethylthio)benzaldehyde ≥98%
4-(Trifluoromethylthio)benzaldehyde

Hazard: Irritant/Air Sensitive/Stench

Formula: C₈H₅F₃OS
Melting Pt: 29-31°C
MDL Number: MFCD00129191
CAS Number: 4021-50-5

Order Now

Specification Test Results

Melting point 29-31°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR