3-(Trifluoromethyl)benzoic acid hydrazide ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(Trifluoromethyl)benzhydrazide, 3-(Trifluoromethyl)benzohydrazide

005421-1G 005421-5G
101788-922EA 70.77 USD
101788-922 101788-924
3-(Trifluoromethyl)benzoic acid hydrazide ≥95%
3-(Trifluoromethyl)benzoic acid hydrazide

Hazard: Irritant

Formula: C₈H₇F₃N₂O
MW: 204.1492 g/mol
Melting Pt: 108-110°C
MDL Number: MFCD00221476
CAS Number: 22227-25-4

Order Now

Specification Test Results

Melting point 108-110°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR