2-(Trifluoromethyl)benzoic acid hydrazide
Supplier: Matrix Scientific
Synonyms:
2-(Trifluoromethyl)benzohydrazide, 2-(Trifluoromethyl)benzhydrazide
005420-1G
005420-5G
101788-918EA
230.77
USD
101788-918
101788-920
2-(Trifluoromethyl)benzoic acid hydrazide
2-(Trifluoromethyl)benzoic acid hydrazide
Hazard: Irritant
Formula:
C₈H₇F₃N₂O MW: 204.150 g/mol Melting Pt: 132-134°C |
MDL Number:
MFCD00221473 CAS Number: 344-95-6 |
Specification Test Results
Melting point | 132-134°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...