4-(Trifluoromethyl)benzamidine hydrochloride dihydrate

Supplier: Matrix Scientific

005419-1G 005419-5G
101788-914EA 47.69 USD
101788-914 101788-916
4-(Trifluoromethyl)benzamidine hydrochloride dihydrate
4-(Trifluoromethyl)benzamidine hydrochloride dihydrate

Hazard: Irritant

Formula: C₈H₁₂ClF₃N₂O₂
Melting Pt: 155 °C
MDL Number: MFCD00122370
CAS Number: 175278-62-3

Order Now

Specification Test Results

Melting point 155°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR