6-(Trifluoromethyl)nicotinic acid ≥98%
Supplier: Matrix Scientific
005417-1G
005417-5G
101788-910EA
15.38
USD
101788-910
101788-912
6-(Trifluoromethyl)nicotinic acid ≥98%
6-(Trifluoromethyl)nicotinic acid
Hazard: Irritant
Formula:
C₇H₄F₃NO₂ MW: 191.11 g/mol Melting Pt: 191-192°C |
MDL Number:
MFCD00792430 CAS Number: 231291-22-8 |
Specification Test Results
Melting point | 191-192°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...