7-(Trifluoromethyl)-4-quinolinol ≥98%

Supplier: Matrix Scientific

005411-5G 005411-1G
101788-894EA 144.62 USD
101788-894 102832-768
7-(Trifluoromethyl)-4-quinolinol ≥98%
7-(Trifluoromethyl)-4-quinolinol

Hazard: Irritant

Formula: C₁₀H₆F₃NO
Melting Pt: 266-269°C
MDL Number: MFCD00006779
CAS Number: 322-97-4

Order Now

Specification Test Results

Melting point 266-269°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR