6-(Trifluoromethyl)-4-quinolinol ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Hydroxy-6-(trifluoromethyl)quinoline

005410-1G 005410-5G
101788-890EA 43.08 USD
101788-890 101788-892
6-(Trifluoromethyl)-4-quinolinol ≥97%
6-(Trifluoromethyl)-4-quinolinol

Hazard: Irritant

Formula: C₁₀H₆F₃NO
MW: 213.16 g/mol
Melting Pt: 265…268 °C
MDL Number: MFCD00209673
CAS Number: 49713-51-1

Order Now

Specification Test Results

Melting point 265-268°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR