2-(Trifluoromethyl)-4-quinolinol ≥95%
Supplier: Matrix Scientific
Synonyms:
2-(Trifluoromethyl)quinolin-4-ol, 4-Hydroxy-2-(trifluoromethyl)quinoline
005409-1G
005409-5G
101788-886EA
13.85
USD
101788-886
101788-888
2-(Trifluoromethyl)-4-quinolinol ≥95%
2-(Trifluoromethyl)-4-quinolinol
Hazard: Irritant
Formula:
C₁₀H₆F₃NO MW: 213.1560 g/mol Melting Pt: 198…203 °C |
MDL Number:
MFCD00153196 CAS Number: 1701-18-4 |
Specification Test Results
Melting point | 198-203°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...