3-(Trifluoromethyl)-2-pyrazolin-5-one ≥97%

Supplier: Matrix Scientific

Synonyms: 3-(Trifluoromethyl)-1H-pyrazol-5(4H)-one

005407-5G 005407-25G
101788-880EA 106.15 USD
101788-880 102550-864
3-(Trifluoromethyl)-2-pyrazolin-5-one ≥97%
3-(Trifluoromethyl)-2-pyrazolin-5-one

Hazard: Irritant

Formula: C₄H₃F₃N₂O
MW: 152.08 g/mol
Melting Pt: 214…217 °C
MDL Number: MFCD00052748
CAS Number: 401-73-0

Order Now

Specification Test Results

Melting point 214-217°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR