4-(Trifluoromethyl)nicotinic acid ≥98%

Supplier: Matrix Scientific

005403-1G 005403-5G
101788-866EA 43.08 USD
101788-866 101788-868
4-(Trifluoromethyl)nicotinic acid ≥98%
4-(Trifluoromethyl)nicotinic acid

Hazard: Irritant

Formula: C₇H₄F₃NO₂
MW: 191.11 g/mol
Melting Pt: 147-149°C
MDL Number: MFCD00082626
CAS Number: 158063-66-2

Order Now

Specification Test Results

Melting point 147-149°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR