5-(Trifluoromethoxy)indole-2-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 5-(Trifluoromethoxy)-1H-indole-2-carboxylic acid

005333-100MG 005333-500MG
101788-682EA 47.69 USD
101788-682 101788-684
5-(Trifluoromethoxy)indole-2-carboxylic acid
5-(Trifluoromethoxy)indole-2-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₆F₃NO₃
MW: 245.16 g/mol
Melting Pt: 199…201 °C
MDL Number: MFCD00276997
CAS Number: 175203-84-6

Order Now

Specification Test Results

Melting point 199-201°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR