4-(Trifluoromethoxy)benzoic acid hydrazide ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(Trifluoromethoxy)benzohydrazide, 4-(Trifluoromethoxy)benzhydrazide

005316-1G 005316-5G
101788-666EA 56.92 USD
101788-666 101788-668
4-(Trifluoromethoxy)benzoic acid hydrazide ≥95%
4-(Trifluoromethoxy)benzoic acid hydrazide

Hazard: Irritant

Formula: C₈H₇F₃N₂O₂
MW: 220.15 g/mol
Melting Pt: 110-120°C
MDL Number: MFCD00204234
CAS Number: 175277-18-6

Order Now

Specification Test Results

Melting point 110-120°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR